Stampa…
Racks and lids for storage 2D barcoded tubes, Matrix™
Fornitore: THERMO MATRIX TECHNOLOGIES
4890 4893 4896 4897 4898 4899 4900 4905 4903 4897-BR 4900-BR 4898-BR
479-2061EA 0 EUR
479-2061 MATR4893 479-2062 MATR4897 MATR4898 479-2063 MATR4900 479-2065 479-2064 MATR4897-BR MATR4900-BR MATR4898-BR
Racks and lids for storage 2D barcoded tubes, Matrix™
Rack Supporti per vials da criogenia
Compatible with automated systems and rack features locking lid to secure samples.
  • Made of plastic
  • Superior storage rack design
  • Latch racks to save precious space
  • Color: white
The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop. The lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications.