Stampa…
Wax plates for sealing capillary tubes
Fornitore: MARIENFELD
2960409
MARI2960409EA 36.7 EUR
MARI2960409
Wax plates for sealing capillary tubes
Guarnizioni Composti sigillanti
Wax plates are used for sealing capillary tubes for micro hematocrits.
  • Two deposit areas on a plastic base
  • Numbered up to 48
Contain modelling clay and additives of food filler material and natural bees wax. We consciously forego adding any chemicals which could affect the plastic material after some time.
Confezione: Packed in poly bags.