Stampa…
Test paper, cyanide
Fornitore: MACHEREY-NAGEL
90604
MANA90604EA 56.2 EUR
MANA90604
Test paper, cyanide
Kit per analisi ambientali
This test paper allows the quick and easy detection of hydrocyanic acid and cyanides in aqueous solutions and extracts. They are for example formed in the manufacture of fruit brandies and are toxic even in low concentrations.
  • Colour change: Pale-green to blue
To detect cyanides in solution, a drop of concentrated sulphuric acid is added to 10 ml sample. Hydrocyanic acid gas is formed and can be detected at the boundary layer between water and air.
Limit of sensitivity: 0,2 mg/L hydrocyanic acid (HCN)
Informazioni per l'ordine: Reel of 5 m length and 10 mm width