Stampa…

3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT) 98%, Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY
Attenzione


M-Clarity™ quality level = MQ200

M2128-5G M2128-100MG M2128-10G M2128-1G M2128-250MG M2128-500MG
M2128-5GEA 441 EUR
M2128-5G M2128-100MG M2128-10G M2128-1G M2128-250MG M2128-500MG
3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT) 98%, Sigma-Aldrich®
3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT)

Thiazolyl Blue Tetrazolium Blue (MTT) may be used in measurement of cell proliferation. MTT produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells. The blue crystals are solubilized with acidified isopropanol and the intensity is measured colorimetrically at 570 nm. MTT has been used as a histochemical/cytochemical reagent and for the detection of NAD. ADP-linked enzyme systems in tissue cannot be detected with MTT, due to binding of the cation by the cyanide trap used. MTT is rapidly reduced to the formazan, which chelates with nickel, copper, and cobalt; the cobalt chelate has been used in oxidative systems.


  • Thiazolyl Blue Tetrazolium Bromide has been used for the following assays:
  • Proliferation assay
  • TNF (tumor necrosis factor) bioassay

Formula: C₁₈H₁₆N₅BrS
Punto di fusione: 195 °C (dec., lit.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561211eb7a01c80370a0145ebbb045631152528d&vid=201dbd708a0c122ed9fb72129893fc2c816b2632&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Purity (HPLC) ≥98.0%
Solubility (Turbidity), c = 5 mg/ml (0.5%); Water Clear
Solubility (Colour) Yellow to Very Dark Yellow and Yellow-Green
Recommended Retest Period 5 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR