Stampa…

AEBSF cloridrato (4 - (2-Aminoethil) benzenesulfonil fluoruro cloridrato) ≥98%, per proteomica

Fornitore: VWR Chemicals
Attenzione

Synonyms: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride

M182-500MGEA 0 EUR
M182-500MG
AEBSF cloridrato (4 - (2-Aminoethil) benzenesulfonil fluoruro cloridrato) ≥98%, per proteomica
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Formula: C₈H₁₁ClFNO₂S
Peso molecolare: 239.69 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00132962
CAS Number: 30827-99-7
UN: 3261
ADR: 8 II

Visualizza i codici

Risultati del test di specificazione

Protease NONE
Purity (Titration) 98 %
Solubility (0.02%, Water) PASS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR