Stampa…
Draining racks
Fornitore: LP ITALIANA
291195
LPIT291195EA 118 EUR
LPIT291195
Draining racks
Rack Rack di Drenaggio
Drying rack, made of PP, with 81 places that can be fully disassembled.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • With carrying handle