Stampa…
pH electrode
Fornitore: Lovibond
721230
LOVI721230EA 179 EUR
LOVI721230
pH electrode
Elettrodi Elettrodi pH
Combined pH/temperature electrode with temperature sensor NTC 30 kOhm and BNC connection.