Stampa…
Test kits, aluminum (Testpak), CHECKIT®
Fornitore: Lovibond
147700
LOVI147700EA 42 EUR
LOVI147700
Test kits, aluminum (Testpak), CHECKIT®
Kit per analisi ambientali
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
Testpak is a simple and cost-effective means of extending the use of an existing CHECKIT® comparator instrument to a new test parameter.
Informazioni per l'ordine: Each Testpak includes the required CHECKIT® disc, tablet reagents (normally for 30 tests), two cells, stirring rod and detailed instructions for the desired method.