Stampa…
Stirrer bearings, KPG®
Fornitore: Lenz Laborglas GmbH & CO.KG
06300010 06300116
LENZ06300010EA 35.4 EUR
LENZ06300010 LENZ06300116
Stirrer bearings, KPG®
Dispositivi agitatori Accessori di agitazione
These stirrer bearings are made of precision calibrated glass tube.
  • Surface is grounded and polished
  • Interchangeable