Stampa…
Tubings, insulated
Fornitore: LAUDA
LZM047 LZM046 LZM048 LZM049 LZM040 LZM041 LZM042 LZM043
LAUDLZM047EA 652 EUR
LAUDLZM047 LAUDLZM046 LAUDLZM048 LAUDLZM049 LAUDLZM040 LAUDLZM041 LAUDLZM042 LAUDLZM043
Tubings, insulated
Tubazioni Tubazione di trasferimento
These metal hoses with thermal insulation consist of flexible corrugated stainless steel tubing and are unaffected by the usual bath liquids. The hose is secured to the connection with a ring nut. The fitting consists of a taper-cone system and not involves any flexible seal.