Stampa…
Heat Transfern Liquid, Ultra 301
Fornitore: LAUDA
LZB353
LAUDLZB353EA 593 EUR
LAUDLZB353
Heat Transfern Liquid, Ultra 301
Liquidi per bagni termostatici
Heat transfer liquid for temperatures up to over 300 °C.
  • Silicone-free liquid, ideal for surface-sensitive applications
  • The safety data sheet must be observed for uses in bath thermostats