Stampa…
Heating and cooling process thermostats, Integral XT
Fornitore: LAUDA
L002673 L002674 L002675 L002676 L002677 L002678 L002684 L002685 L002687 L002689 L002683 L002681
LAUDL002673EA 0 EUR
LAUDL002673 LAUDL002674 LAUDL002675 LAUDL002676 LAUDL002677 LAUDL002678 LAUDL002684 LAUDL002685 LAUDL002687 LAUDL002689 LAUDL002683 LAUDL002681
Heating and cooling process thermostats, Integral XT
Termostati a circolazione Sistemi a circolazione chiusa Sistemi di controllo della temperatura
Process thermostats with integrated cooling system for dynamic temperature control within external circuits.
  • Coloured TFT display for simultaneous indication of actual and set values and graphic illustration of the temperature profile
  • Clear text menu navigation, six selectable languages DE, EN, FR, ES, IT and RU
  • Management of heat transfer liquids with stored properties
  • Easy input via cursor and soft keys. Additional Tmax key for overtemperature protection
  • SelfCheck Assistant for system diagnosis
  • Fully electronic continuous controller with PID action for