Stampa…
Super drum, omologated
Fornitore: LAMAPLAST
3209
LAMA3209EA 1340 EUR
LAMA3209
Super drum, omologated
Botti, fusti, tamburi
These hermetic safety container are produced by rotational system in high density polyethylene with an high molecular weight. Suitable for the transport and storing of metal drums containing harmful, toxic substances.
  • Resistant to impact, chemical and atmospheric agents
  • Double wall lid with dutral seal and stainless steel iron ring for a perfect hermetic seal