Stampa…

4-Amino-1-boc-piperidine-4-carboxylic acid 98%

Fornitore: Thermo Fisher Scientific

L19794.03
L19794.03EA 167 EUR
L19794.03
4-Amino-1-boc-piperidine-4-carboxylic acid 98%
4-Amino-1-boc-piperidine-4-carboxylic acid

Purity: 98% CAS No.: 183673-71-4 Molecular Formula: C11H20N2O4 Formula Weight: 244.29 Storage Temperature: 2°C to 8°C Melting Point: ca 290° dec. Beilstein Reference: 7588844 MDL No.: MFCD01318728

Formula: C₁₁H₂₀N₂O₄
Punto di fusione: ∼ 290 °C (dec.)
MDL Number: MFCD01318728
CAS Number: 183673-71-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR