Stampa…

Isobutyl-4-hydroxybenzoate ≥98%

Fornitore: Thermo Fisher Scientific

Synonyms: Isobutyl 4-hydroxybenzoate, Isobutylparaben

L17587.06 L17587.18
L17587.06EA 30.2 EUR
L17587.06 L17587.18
Isobutyl-4-hydroxybenzoate ≥98%
Isobutyl-4-hydroxybenzoate

Purity: 98% CAS No.: 4247-02-3 Molecular Formula: C11H14O3 Formula Weight: 194.23 Melting Point: 74-78° Beilstein Reference: 2642305

Formula: C₁₁H₁₄O₃
Punto di fusione: 74…78 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00020167
CAS Number: 4247-02-3
EINECS: 224-208-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR