Stampa…

4-Hydroxy-6-methyl-2-pyrone 98%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: 4-Hydroxy-6-methyl-2H-pyran-2-one, 3,5-Dihydroxysorbic acid d-lactone, 2-hydroxy-6-methyl-pyran-4-one

L11457.14 L11457.22
L11457.14EA 59.7 EUR
L11457.14 L11457.22
4-Hydroxy-6-methyl-2-pyrone 98%
4-Hydroxy-6-methyl-2-pyrone

Purity: 98% CAS No.: 675-10-5 Molecular Formula: C6H6O3 Formula Weight: 126.11 Melting Point: ca 186° dec. Beilstein Reference: 113815 MDL No.: MFCD00006641

Formula: C₆H₆O₃
Punto di fusione: ∼ 186 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006641
CAS Number: 675-10-5
EINECS: 211-619-2

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR