Stampa…

(S)-(-)-1,1'-Bi-2-naphthol ≥99%

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol

L08423.03 L08423.06 L08423.14
L08423.03EA 28.5 EUR
L08423.03 L08423.06 L08423.14
(S)-(-)-1,1'-Bi-2-naphthol ≥99%
(S)-(-)-1,1'-Bi-2-naphthol

Purity: 99% CAS No.: 18531-99-2 Molecular Formula: C20H14O2 Formula Weight: 286.33 Odour: Characteristic Melting Point: 206-211° Beilstein Reference: 3592999 Merck Reference: 14,1226 Optical Rotation: -34.5 (c=1 in THF) UN No.: 2811 MDL No.: MFCD00004068

Formula: C₂₀H₁₄O₂
Peso molecolare: 286.33 g/mol
Punto di fusione: 206…211 °C
Densità: 1.301
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD19441803
CAS Number: 18531-99-2
UN: 2811
ADR: 6.1 III
Merck Index: 14,1226

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR