Stampa…

4-(2-Chloroethyl)morpholine hydrochloride ≥99%

Fornitore: Thermo Fisher Scientific
Pericolo

L05771.22 L05771.36
L05771.22EA 33.8 EUR
L05771.22 L05771.36
4-(2-Chloroethyl)morpholine hydrochloride ≥99%
4-(2-Chloroethyl)morpholine hydrochloride

Purity: 99% CAS No.: 3647-69-6 Molecular Formula: C6H12ClNO.HCl Formula Weight: 186.08 Melting Point: 184-186° Beilstein Reference: 3684083 UN No.: 2923 MDL No.: MFCD00012797

Formula: C₆H₁₃Cl₂NO
Punto di fusione: 184…186 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012797
CAS Number: 3647-69-6
EINECS: 222-881-2
UN: 2923
ADR: 8,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR