Stampa…

Setaccio molecolare 4A (0,4 nm, 4 Å) 3 - 5 mm (0.12-0.20 in)

Fornitore: Thermo Fisher Scientific

Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate

L05466.30 L05466.0B L05466.0I
L05466.30EA 44.5 EUR
L05466.30 L05466.0B L05466.0I
Setaccio molecolare 4A (0,4 nm, 4 Å) 3 - 5 mm (0.12-0.20 in)
Setaccio molecolare 4A (0,4 nm, 4 Å)

CAS No.: 70955-01-0 MDL No.: MFCD00131613

Densità: 1,1…2.2 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR