Stampa…
Cell culture media, RPMI 1640
Fornitore: Biowest
L0490-500 L0492-500 L0495-500 L0496-500 L0498-500 L0500-100 L0500-500 L0501-100 L0501-500 P0860-N1L P0860-N10L P0870-N1L P0870-N10L P0871-N1L P0880-N10L P0883-N1L P0870-N50L P0876-N1L P0876-N10L P0876-N50L P0880-N50L P0883-N10L P0860-N5L P0860-N50L P0880-N1L P0871-N10L L0498-100 L0503-500 L0505-500 L0501-1000
L0490-500EA 12 EUR
L0490-500 L0492-500 L0495-500 L0496-500 L0498-500 L0500-100 L0500-500 L0501-100 L0501-500 P0860-N1L P0860-N10L P0870-N1L P0870-N10L P0871-N1L P0880-N10L P0883-N1L P0870-N50L P0876-N1L P0876-N10L P0876-N50L P0880-N50L P0883-N10L P0860-N5L P0860-N50L P0880-N1L P0871-N10L L0498-100 L0503-500 L0505-500 L0501-1000
Cell culture media, RPMI 1640
Terreni per colture cellulari
RPMI are general purpose enriched media with extensive applications for a broad spectrum of mammalian and hybridoma cells, including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes. It was originally formulated for suspension cultures and monolayer culture of human leukaemia cells.