Stampa…

4-(2-Chloroethyl)benzoic acid 97%

Fornitore: Thermo Fisher Scientific
Attenzione

L04068.09 L04068.18
L04068.09EA 96.9 EUR
L04068.09 L04068.18
4-(2-Chloroethyl)benzoic acid 97%
4-(2-Chloroethyl)benzoic acid

Purity: 97% CAS No.: 20849-78-9 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 187-189° Beilstein Reference: 2084041 MDL No.: MFCD00013995

Formula: C₉H₉ClO₂
Punto di fusione: 187…189 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013995
CAS Number: 20849-78-9
EINECS: 244-080-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR