Stampa…

Solfato di bis (8-idrossichinolinio) monoidrato ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Quinolin-8-ol sulphate hydrate, Quinolin-8-ol hemisulphate hemihydrate, 8-Hydroxyquinoline sulphate monohydrate, 8-Hydroxyquinolinium hemisulphate hemihydrate, 8-Hydroxyquinoline hemisulphate salt hemihydrate

L02743.14 L02743.22
L02743.14EA 25.4 EUR
L02743.14 L02743.22
Solfato di bis (8-idrossichinolinio) monoidrato ≥99%
Solfato di bis (8-idrossichinolinio) monoidrato

Purity: 99% CAS No.: 207386-91-2 Molecular Formula: C18H14N2O2.H2SO4.H2O Formula Weight: 406.42 (388.40anhy) Melting Point: 176-179° Beilstein Reference: 3760550

Formula: C₁₈H₁₆N₂O₆S·1H₂O
Punto di fusione: 176…179 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149610
CAS Number: 207386-91-2
EINECS: 205-137-1

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR