Stampa…

(Cyclopropylmethyl)triphenylphosphonium bromide ≥98%

Fornitore: Thermo Fisher Scientific
Attenzione

L01145.06 L01145.14
L01145.06EA 96.9 EUR
L01145.06 L01145.14
(Cyclopropylmethyl)triphenylphosphonium bromide ≥98%
(Cyclopropylmethyl)triphenylphosphonium bromide

Purity: 98+% CAS No.: 14799-82-7 Molecular Formula: C22H22BrP Formula Weight: 397.29 Melting Point: 178-182° Beilstein Reference: 4068302

Formula: C₁₄H₂₉ClNP
Punto di fusione: 178…182 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00051878
CAS Number: 14799-82-7
EINECS: 238-862-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR