Stampa…

Penicillin : Streptomycin soluzione 6,0/10,0 g/L 100X

Fornitore: Biowest

L0022-100 L0022-020
L0022-100EA 17 EUR
L0022-100 L0022-020
Penicillin : Streptomycin soluzione 6,0/10,0 g/L 100X
Penicillin/Streptomycin

Penicillin-Streptomycin acts against Gram-negative and Gram-positive bacteria.


  • Store at –20 °C
  • pH of 6±1
  • Shelf life of 18 months


Endotoxin level at 10 ml/l of solution is <0,1 EU/ml.


Attenzione: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

CAS Number: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR