Stampa…
Cooling racks
Fornitore: KOEHLER TECHNISCHE PRODUKTEN
A-1070
KOEHA-1070EA 174 EUR
KOEHA-1070
Cooling racks
Rack Rack di Raffreddamento
PCR vials may be kept at a temperature of 0 °C for at least 30 min if the cooler is cooled down to –20 °C for approximately 12 h.