Stampa…
Foam soaps, KLEENEX®
Fornitore: KIMBERLY CLARK
6345
KIMB6345EA 158 EUR
KIMB6345
Foam soaps, KLEENEX®
Cura della pelle Saponi e detergenti per la pelle
This clear foaming hand soap improves personal hygiene. Ideal for using in busy hygiene critical washrooms, gentle cleaning, without irritating the skin.
  • Dispenses an ideal amount of soap for each wash when hands are washed frequently
  • Prevents overuse and delivers up to twice as many washes per litre as conventional liquid soaps
  • Colourant-free and fragrance-free
Informazioni per l'ordine: 1,2 litre electronic cassettes for use with the KIMBERLY-CLARK PROFESSIONAL* Touchless Electronic Skincare Dispenser; 1.5 litre bottles for use with the KIMBERLY-More Product Information