Stampa…
Pipette tips
Fornitore: KARTELL
961 963 965 967 968 969 9640 9641 958 9643 957 9646 9645 935 9644 9647 9648 9650 9657 971 979 984 9847 9848 9585 9605 9615 9745 9746 9747 9624 9748 9749 9751 9752 9753 9754 940 941 942 943 964 0095600 0095900 0096006 0098006 0964900
612-1418EA 18.1 EUR
612-1418 KART963 613-0500 KART967 KART968 KART969 KART9640 KART9641 KART958 KART9643 KART957 KART9646 KART9645 KART935 KART9644 KART9647 KART9648 KART9650 KART9657 KART971 KART979 KART984 KART9847 KART9848 KART9585 KART9605 KART9615 KART9745 KART9746 KART9747 KART9624 KART9748 KART9749 KART9751 KART9752 KART9753 KART9754 KART940 KART941 KART942 KART943 KART964 KART0095600 KART0095900 KART0096006 KART0098006 KART0964900
Pipette tips
Puntali per pipette
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • Tips in racks come with snap-on lid
  • Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
  • Containers and tips