Stampa…
Becher, PTFE
Fornitore: KARTELL
720 721 723 717 718
KART720EA 84.7 EUR
KART720 KART721 KART723 KART717 KART718
Becher, PTFE
Becher
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +250 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.