Stampa…
Hand protectors, Hot Grip
Fornitore: KARTELL
3915
KART3915EA 40.8 EUR
KART3915
Hand protectors, Hot Grip
Protezioni termiche per le mani
These hand protectors made of silicone rubber provide protection against hot containers up to +260 °C and cold containers down to –57 °C. Internal relief surface.