Stampa…
Cuvettes for Fibrintimer®
Fornitore: KARTELL
2630
KART2630EA 48.8 EUR
KART2630
Cuvettes for Fibrintimer®
Cuvette Cuvette speciali
Polystyrene cuvettes for Fibrintimer used to determine the coagulation-time of blood.
Certificazioni: IVD (in vitro diagnostic) certified.
Informazioni per l'ordine: Supplied with 500 metallic mixers.