Stampa…
Screw caps, for DIN 168 bottles, plastilab®
Fornitore: KARTELL
15950 15990 16030 16250 16290
KART15950EA 156 EUR
KART15950 KART15990 KART16030 KART16250 KART16290
Screw caps, for DIN 168 bottles, plastilab®
Ghiere Tappi per bottiglie
Screw caps made of PP, it has cap tag to avoid sample contamination in transit.
Standard screw caps for polyethylene bottles manufactured to DIN 168.