Stampa…
Safety spectacle, Galactus™
Fornitore: JSP
ASA480-0A7-600
JSPAASA480-0A7-600EA 23.8 EUR
JSPAASA480-0A7-600
Safety spectacle, Galactus™
Occhiali
Adaptable and ultra-light safety spectacle with base ten lens.
  • Stylish premium safety eyewear
  • Ultra-light TPR frame
  • Adaptable nose bridge
  • Base ten lens
  • Soft touch arms
Galactus™ premium safety eyewear features a base 10 curved lens which follows facial contours perfectly, a unique mouldable nose bridge system that can be adjusted to accommodate all high/low/thin or wide nose bridge profiles, and a frame that utilises a fusion of polymer and TPE to be both lightweight and super strong.
Soft touch arms with elastomer-injected sleeve for more comfortMore Product Information