Stampa…

Pectin (from citrus)

Fornitore: Thermo Fisher Scientific

J61021.22 J61021.36
J61021.22EA 40.4 EUR
J61021.22 J61021.36
Pectin (from citrus)
Pectin

CAS No.: 9000-69-5 Physical Form: Powder Appearance: Yellow to pale brown MDL No.: MFCD00081838

Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00081838
CAS Number: 9000-69-5
EINECS: 232-553-0
Merck Index: 14,7063

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR