Stampa…

Ponceau S per elettroforesi

Fornitore: Thermo Fisher Scientific
Attenzione

J60744.09 J60744.30 J60744.18
J60744.09EA 21.3 EUR
J60744.09 J60744.30 J60744.18
Ponceau S per elettroforesi
Ponceau S

CAS No.: 6226-79-5 Molecular Formula: C22H12N4Na4O13S4 Formula Weight: 760.58 Physical Form: Powder Appearance: Red-brown


Protein stain for electrophoresis

Formula: C₂₂H₁₂N₄Na₄O₁₃S₄
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003892
CAS Number: 6226-79-5
EINECS: 228-319-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR