Stampa…

POPSO, 0.2 M buffer solution, pH 8.0

Fornitore: Thermo Fisher Scientific

J60085.AK
J60085.AKEA 76.8 EUR
J60085.AK
POPSO, 0.2 M buffer solution, pH 8.0
Soluzioni tampone Tamponi biologici

CAS No.: 108321-07-9
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
MDL No.: MFCD00070005

Visualizza i codici

Specifiche

Environmentally Preferable
Forma fisica Liquid
CAS No. 108321-07-9
N. MDL MFCD00070005
Temperatura di conservazione 2°C to 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR