Stampa…
Recirculating chillers, RC 2
Fornitore: IKA
4171000 4173000
IKAA4171000EA 4670 EUR
IKAA4171000 IKAA4173000
Recirculating chillers, RC 2
Termostati a circolazione Sistemi a circolazione chiusa Termostati a circolazione raffreddanti
Designed for cooling applications up to −20 °C with a powerful 400 W cooling capacity, these recirculating chillers are highly efficient and provide stable temperatures. With a wireless controller, it is possible to operate the RC 2 safely and comfortably irrespective of where the device is located.
  • A feed hopper bleed valve enables safe and clean handling of the thermofluid
  • Easy to open front cover allows the air filter to be conveniently cleaned
  • It is possible to measure and control temperature directly in the working medium via the external temperature