Stampa…
Magnetic hotplate stirrer, IKA Plate RCT digital
Fornitore: IKA
25004601
IKAA25004601EA 1172 EUR
IKAA25004601
Magnetic hotplate stirrer, IKA Plate RCT digital
Dispositivi agitatori Agitatori magnetici con piastra riscaldante
The IKA Plate (RCT digital) magnetic hotplate stirrer has an aluminium alloy plate, suitable for optimum heat transfer. It features a glass housing surface for optimum cleaning and chemical resistance. The unit features a strong motor for a high range of speeds, integrated timer and counter function supports the monitoring of sensitive chemical reactions.
  • SmartTemp® hot surface warning to protect against burns
  • Connector for electronic temperature control (via optional PT1000 probe)
  • Regulation of the plate can be optimised by regular firmware updates