Stampa…
Electronic overhead stirrer, NANOSTAR 7.5 digital
Fornitore: IKA
25004356
IKAA25004356EA 986 EUR
IKAA25004356
Electronic overhead stirrer, NANOSTAR 7.5 digital
Dispositivi agitatori Agitatori ad asta
Small and space saving overhead stirerr with great perfomance.
  • Guarantees a constant torque over the entire rpm range from 50 to 2000 min⁻¹
  • Easy to operate using a stable rotary knob
  • Display clearly shows the rotational speed
  • Regular software updates can be undertaken rapidly and simply via the USB interface
The NANOSTAR 7.5 digital overhead stirrer is 30% smaller than the MICROSTAR 7.5 digital and therefore even more space-saving. However, its performance remains the same. NANOSTAR 7.5 digital convinces with perfect basic functionalities, its compactMore Product Information