Stampa…
Immersion Circulators, ICC 150 lite
Fornitore: IKA
20105581
IKAA20105581EA 1080 EUR
IKAA20105581
Immersion Circulators, ICC 150 lite
Termostati a circolazione Sistemi a circolazione aperta Sistemi di controllo della termostatazione
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation. ICC 150 lite is suitable for tempering liquids up to 150 °C. The integrated pressure/suction pump allows the device to be used for internal and external applications.
Increased safety: ICC 150 lite is approved for use with flammable liquids in accordance with DIN 12876, safety class III. It detects an overflow, e.g. due to an unintentional increase in the volume of oils, and switches the heating off safely.