Stampa…
Decontamination solution, RNase ERASE™, MP Biomedicals
Fornitore: MP Biomedicals
112440204 04821682 04821683
ICNA112440204EA 122 EUR
ICNA112440204 ICNA04821682 ICNA04821683
Decontamination solution, RNase ERASE™, MP Biomedicals
Soluzioni detergenti Decontaminanti
RNase ERASE™ decontamination solution has been formulated using three ingredients known to be active against RNase.
  • Removes RNase contamination from glass, plastic, pipettes and equipment
  • After thorough rinsing leaves no residues that are inhibitory to enzymatic reactions
Note: RNase ERASE™ is not compatible with corrodible metal surfaces.
Visualizza i codici
Specifiche
Environmentally Preferable
Condizioni di conservazione Ambient