Stampa…

D-(+)-Proline ≥99%, white powder

Fornitore: MP Biomedicals

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

02195456.2 0219545601 0219545605
ICNA02195456.2EA 40.5 EUR
ICNA02195456.2 ICNA0219545601 ICNA0219545605
D-(+)-Proline ≥99%, white powder
D-(+)-Proline

Soluble in water (5% - clear colourless to very faint yellow solution); slightly soluble in acetone, ethanol.


Soluble in water (5% - clear colourless to very faint yellow solution); slightly soluble in acetone, ethanol.

Formula: C₅H₉NO₂
Peso molecolare: 115.13 g/mol
Punto di fusione: 225…235 °C
MDL Number: MFCD00064317
CAS Number: 344-25-2
EINECS: 206-452-7

Visualizza i codici

Risultati del test di specificazione

Optical Rotation +84,5° to +86,5° (c=4, H2O)
Purity ≥99%
FTIR Conforms to standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR