Stampa…

Reagente di Folin-Ciocalteu per l'analisi dei fenoli

Fornitore: MP Biomedicals
Pericolo

0219518690
ICNA0219518690EA 141 EUR
ICNA0219518690
Reagente di Folin-Ciocalteu per l'analisi dei fenoli
Reattivo di Folin-Ciocalteu

Folin & Ciocalteu`s Phenol Reagent is suitable for determination of total protein by Lowry method. It is a mixture of phosphomolybdate and phosphotungstate used for the colorimetric assay of phenolic and polyphenolic antioxidants.


  • 2N Concentration
  • Used in the Lowry method for determining protein concentration
  • Spray reagent in chromatographic procedures


A procedure for preparation of the Folin & Ciocalteu's phenol reagent has been published. Dissolve 10 g of sodium tungstate and 2.5 g of sodium molybdate in 70 ml of water. Add 5 ml of 85% phosphoric acid and 10 ml of concentrated hydrochloric acid. Reflux for 10 hours. Add 15 g of lithium sulfate, 5 ml of water, and 1 drop of bromine. Reflux for 15 minutes. Cool to room temperature and bring to 100 ml with water.

MDL Number: MFCD00132625
UN: 3264
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR