Stampa…

D(+)-Glucosio, white powder, Molecular Biology Grade

Fornitore: MP Biomedicals

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

0219402483 0219402491
ICNA0219402483EA 27 EUR
ICNA0219402483 ICNA0219402491
D(+)-Glucosio, white powder, Molecular Biology Grade
D(+)-Glucosio

D-(+)-Glucose is ideal as a culture media component.


Attenzione: Research use only (RUO).

Formula: C₆H₁₂O₆
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
FTIR Conforms to Standard
Optical Rotation (c=10, Water, NH4OH) +52,5 ± 5°
Moisture Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR