Stampa…
Bovine growth hormone (from Pituitary glands), MP Biomedicals
Fornitore: MP Biomedicals
02160074.1
ICNA02160074.1EA 206 EUR
ICNA02160074.1
Bovine growth hormone (from Pituitary glands), MP Biomedicals
Proteine e Peptidi
Bovine growth hormone is a peptide hormone, it is used in regulating metabolic processes.
Biological activity: In responsive cell lines, biological activity is observed at concentrations of 10 to 100 ng/ml.