Stampa…

Sulphamethazine sodium salt, white powder

Fornitore: MP Biomedicals
Attenzione

0215670983 0215670980
ICNA0215670983EA 124 EUR
ICNA0215670983 ICNA0215670980
Sulphamethazine sodium salt, white powder
Sulphamethazine sodium salt

Sulfamethazine is an antibiotic used to clinically treat bronchitis, prostatitis and urinary tract infections. It is used in disposition and depletion kinetic studies. It is used to develop detection techniques for quantification in fluids such as cows’ milk, honey and swine urine.
Sulfamethazine is an antimicrobial sulfur drug that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Sulfamethazine is a competitive inhibitor of bacterial para-aminobenzoic acid (PABA), which is required for bacterial synthesis of folic acid. It induces CYP3A4 expression and is acetylated by N-acetyltransferase. It exhibits sex dependent pharmacokinetics, metabolized by the male specific isoform CYP2C11. Sulfamethazine is bacteriostatic.


  • Presentation: White Powder
  • H2O: soluble (50 mg/mL)
  • Storage temperature: Store at +4 °C.


Storage temperature.: +4 °C

Formula: C₁₂H₁₃N₄NaO₂S
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00068333
CAS Number: 1981-58-4

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR