Stampa…

Piombo tetra acetato ≥95%, in polvere

Fornitore: MP Biomedicals
Pericolo

Synonyms: Lead(IV) tetraacetate, Triacetyloxyplumbyl acetate, Acetic acid lead (IV) salt, Lead (IV) acetate, LTA

0215518225 0215518290 0215518280
ICNA0215518225EA 62.9 EUR
ICNA0215518225 ICNA0215518290 ICNA0215518280
Piombo tetra acetato ≥95%, in polvere
Piombo tetra acetato

Lead tetraacetate smoothly induces the addition of difluorodiiodomethane to alkenes and alkynes.

Formula: (H₃CCOO)₄Pb
Punto di fusione: 175…180 °C
Densità: ∼2,22 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008693
CAS Number: 546-67-8
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III
Merck Index: 13,05442

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR