Stampa…

Rame (II) nitrato triidrato, in polvere

Fornitore: MP Biomedicals

0215350180 0215350190
ICNA0215350180EA 34.5 EUR
ICNA0215350180 ICNA0215350190
Rame (II) nitrato triidrato, in polvere
Rame (II) nitrato triidrato
Formula: Cu(NO₃)₂·3H₂O
MDL Number: MFCD00149671
CAS Number: 10031-43-3
UN: 1477
ADR: 5.1,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR