Stampa…

Acido Nitrilotriacetico ≥98%, white powder ACS

Fornitore: MP Biomedicals
Attenzione

0215255190 0215255191 0215255180
ICNA0215255190EA 0 EUR
ICNA0215255190 ICNA0215255191 ICNA0215255180
Acido Nitrilotriacetico ≥98%, white powder ACS
Acido Nitrilotriacetico

Nitrilotriacetic acid is an excellent chelating agent which sequester metal ions to form water-soluble complexes; It has many industrial applications. Because of its ability to chelate calcium and magnesium ions.


Nitrilotriacetic acid is an excellent chelating agent which sequester metal ions to form water-soluble complexes; It has many industrial applications. Because of its ability to chelate calcium and magnesium ions.


Soluble in 1M Sodium Hydroxide (100 mg/ml).

Formula: C₆H₉NO₆
Punto di ebollizione: 498 °C (1013 hPa)
Punto di fusione: 242 °C
Densità: 1,67 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004287
CAS Number: 139-13-9
EINECS: 205-355-7
UN: 2923
ADR: 8,II
Merck Index: 12,06675

Visualizza i codici

Risultati del test di specificazione

Assay ≥98%
Clarity of solution Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR