Stampa…

NADPH-Na4 ≥95%

Fornitore: MP Biomedicals

0215174225 0215174250 0215174280 0215174283 0215174290 0215174291
ICNA0215174225EA 119 EUR
ICNA0215174225 ICNA0215174250 ICNA0215174280 ICNA0215174283 ICNA0215174290 ICNA0215174291
NADPH-Na4 ≥95%
NADPH-Na4

50 mg/ml, clear colourless to slightly yellow solution.

Formula: C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number: MFCD00036263
CAS Number: 2646-71-1

Visualizza i codici

Risultati del test di specificazione

Synonyms β-NADPH; TPNH; Coenzyme II reduced form tetrasodium salt; Dihydronicotinamide-adenine dinucleotide phosphate tetrasodium salt; Triphosphopyridine nucleotide reduced tetrasodium salt
Form White Powder
Storage Store at -0 °C, Desiccate, Protect from light, Store under nitrogen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR