Stampa…

(4-Nitrophenyl)-α-D-galactopyranoside, in polvere

Fornitore: MP Biomedicals

0210247105 02102471.2 02102471.5 0210247101
ICNA0210247105EA 812 EUR
ICNA0210247105 ICNA02102471.2 ICNA02102471.5 ICNA0210247101
(4-Nitrophenyl)-α-D-galactopyranoside, in polvere
(4-Nitrophenyl)-α-D-galactopyranoside

p-Nitrophenyl-alpha-D-galactopyranoside is used as a substrate for α-galactosidase.


Soluble in water (5% w/v), HCl (50 mg/ml - clear, colourless solution) & methanol (100 mg/ml).

Formula: C₁₂H₁₅NO₈
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00065050
CAS Number: 7493-95-0
EINECS: 231-343-6

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
FTIR Conforms to Standard C
Melting Point 164 - 176°C
Optical Rotation (c=1, 1N HCl) +240° ± 20°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR